Assay Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011186.1 | NP_035316.1 | 2 - 3 | 510 | 63 | |
| GenBank mRNA | AB003304.1 | - | 2 - 3 | 513 | 63 | |
| AF060091.1 | - | 2 - 3 | 510 | 63 | ||
| AK010441.1 | - | 2 - 3 | 530 | 63 | ||
| AK133839.1 | - | 2 - 3 | 531 | 63 | ||
| BC012246.1 | - | 2 - 3 | 518 | 4167355 | 63 | |
| BC106144.1 | - | 2 - 3 | 513 | 4953887 | 63 | |
More Information
Set Membership: |
|

Psmb5
Genomic Map