Assay Details
| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011240.3 | NP_035370.2 | 27 - 28 | 8649 | 69 | |
| GenBank mRNA | BC030915.1 | - | 2 - 3 | 364 | 69 | |
More Information
Set Membership: |
|

Ranbp2
Genomic Map