Assay Details
| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011240.3 | NP_035370.2 | 28 - 29 | 8982 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ranbp2| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011240.3 | NP_035370.2 | 28 - 29 | 8982 | 102 | |
Set Membership: |
|