Assay Details
| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011240.3 | NP_035370.2 | 3 - 4 | 382 | 119 | |
| GenBank mRNA | AK040322.1 | - | 3 - 4 | 359 | 119 | |
| AK041932.1 | - | 3 - 4 | 315 | 119 | ||
| AK145306.1 | - | 3 - 4 | 313 | 119 | ||
More Information
Set Membership: |
|

Ranbp2
Genomic Map