Assay Details
| Gene Symbol: Ranbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011240.3 | NP_035370.2 | 4 - 5 | 523 | 123 | |
| GenBank mRNA | AK040322.1 | - | 4 - 5 | 500 | 123 | |
| AK041932.1 | - | 4 - 5 | 456 | 123 | ||
| AK145306.1 | - | 4 - 5 | 454 | 123 | ||
More Information
Set Membership: |
|

Ranbp2
Genomic Map