Assay Details
| Gene Symbol: Rps12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011295.6 | NP_035425.2 | 2 - 3 | 105 | 107 | |
| GenBank mRNA | AK002848.1 | - | 2 - 3 | 97 | 107 | |
| AK008057.1 | - | 2 - 3 | 97 | 107 | ||
| AK032757.1 | - | 2 - 3 | 97 | 107 | ||
| AK132208.1 | - | 2 - 3 | 98 | 107 | ||
| AK166923.1 | - | 2 - 3 | 95 | 107 | ||
| AK166992.1 | - | 2 - 3 | 100 | 107 | ||
| AK168324.1 | - | 2 - 3 | 97 | 107 | ||
| AK169215.1 | - | 2 - 3 | 97 | 107 | ||
| BC002079.1 | - | 2 - 3 | 81 | 3489757 | 107 | |
| BC089338.1 | - | 2 - 3 | 93 | 6407567 | 107 | |
| BC089339.1 | - | 2 - 3 | 79 | 6408026 | 107 | |
| BC090257.1 | - | 2 - 3 | 72 | 4209024 | 107 | |
| BC092044.1 | - | 2 - 3 | 79 | 2655660 | 107 | |
| BC096653.1 | - | 2 - 3 | 108 | 1528457 | 107 | |
| BC099377.1 | - | 2 - 3 | 81 | 30936226 | 107 | |
| BY128264.1 | - | 2 - 3 | 107 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002848.1, AK008057.1, AK032757.1, AK166923.1, AK166992.1, AK168324.1, AK169215.1, BC002079.1, BC089338.1, BC089339.1, BC090257.1, BC092044.1, BC096653.1, BC099377.1, NM_011295.6 transcripts.
Set Membership: |
|

Rps12
Genomic Map