Assay Details
| Gene Symbol: Sprr2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011474.3 | NP_035604.1 | 1 - 2 | 45 | 69 | |
| GenBank mRNA | AY158992.1 | - | 1 - 2 | 45 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY158992.1, NM_011474.3 transcripts.
Set Membership: |
|

Sprr2h
Genomic Map