Assay Details
| Gene Symbol: Fxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011814.2 | NP_035944.2 | 10 - 11 | 1366 | 73 | |
| GenBank mRNA | AB025269.1 | - | 9 - 10 | 933 | 73 | |
| AK154243.1 | - | 10 - 11 | 1365 | 73 | ||
| AK172042.1 | - | 10 - 11 | 1379 | 73 | ||
| AK208654.1 | - | 2 - 3 | 176 | 73 | ||
| AK210574.1 | - | 3 - 4 | 246 | 73 | ||
| AK216106.1 | - | 2 - 3 | 172 | 73 | ||
| AK219454.1 | - | 2 - 3 | 163 | 73 | ||
| BC057007.1 | - | 10 - 11 | 1261 | 6816943 | 73 | |
| BC062971.1 | - | 10 - 11 | 1366 | 6415619 | 73 | |
More Information
Set Membership: |
|

Fxr2
Genomic Map