Assay Details
| Gene Symbol: Fxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011814.2 | NP_035944.2 | 14 - 15 | 2082 | 75 | |
| GenBank mRNA | AK154243.1 | - | 14 - 15 | 2081 | 75 | |
| BC057007.1 | - | 14 - 15 | 1977 | 6816943 | 75 | |
| BC062971.1 | - | 14 - 15 | 2082 | 6415619 | 75 | |
More Information
Set Membership: |
|

Fxr2
Genomic Map