Assay Details
| Gene Symbol: Fxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011814.2 | NP_035944.2 | 4 - 5 | 638 | 87 | |
| GenBank mRNA | AB025269.1 | - | 3 - 4 | 205 | 87 | |
| AK154243.1 | - | 4 - 5 | 637 | 87 | ||
| AK172042.1 | - | 4 - 5 | 651 | 87 | ||
| BC057007.1 | - | 4 - 5 | 533 | 6816943 | 87 | |
| BC062971.1 | - | 4 - 5 | 638 | 6415619 | 87 | |
More Information
Set Membership: |
|

Fxr2
Genomic Map