Assay Details
| Gene Symbol: Mpdu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011900.4 | NP_036030.2 | 3 - 4 | 377 | 60 | |
| GenBank mRNA | AB024713.1 | - | 3 - 4 | 305 | 60 | |
| AK040432.1 | - | 3 - 4 | 316 | 60 | ||
| AK080233.1 | - | 3 - 4 | 371 | 60 | ||
| AK089426.1 | - | 3 - 4 | 319 | 60 | ||
| BC026776.1 | - | 3 - 4 | 299 | 4159355 | 60 | |
| BC048871.1 | - | 3 - 4 | 305 | 5364408 | 60 | |
More Information
Set Membership: |
|

Mpdu1
Genomic Map