Assay Details
| Gene Symbol: Cops3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011991.1 | NP_036121.1 | 2 - 3 | 226 | 97 | |
| GenBank mRNA | AF071313.1 | - | 2 - 3 | 226 | 97 | |
| AK048906.1 | - | 2 - 3 | 216 | 97 | ||
| AK142318.1 | - | 2 - 3 | 212 | 97 | ||
| BC068179.1 | - | 2 - 3 | 226 | 30633689 | 97 | |
More Information
Set Membership: |
|

Cops3
Genomic Map