Assay Details
| Gene Symbol: Cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013494.3 | NP_038522.2 | 1 - 2 | 401 | 107 | |
| GenBank mRNA | AK002935.1 | - | 1 - 2 | 46 | 107 | |
| AK013509.1 | - | 1 - 2 | 98 | 107 | ||
| AK047891.1 | - | 1 - 2 | 392 | 107 | ||
| AK160097.1 | - | 1 - 2 | 416 | 107 | ||
| BC010197.1 | - | 1 - 2 | 401 | 3157558 | 107 | |
More Information
Set Membership: |
|

Cpe
Genomic Map