Assay Details
Gene Symbol: H2-Ke6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013543.2 | NP_038571.2 | 1 - 2 | 57 | 77 | |
GenBank mRNA | BC086927.1 | - | 1 - 2 | 57 | 6775958 | 77 |
KJ650217.1 | - | 1 - 2 | 49 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
