Assay Details
| Gene Symbol: Nup50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016714.2 | NP_057923.2 | 1 - 2 | 218 | 62 | |
| GenBank mRNA | AK087706.1 | - | 1 - 2 | 236 | 62 | |
| AK133562.1 | - | 1 - 2 | 253 | 62 | ||
| AK141750.1 | - | 1 - 2 | 258 | 63 | ||
| AK151947.1 | - | 1 - 2 | 223 | 62 | ||
| AK152309.1 | - | 1 - 2 | 223 | 62 | ||
| AK161713.1 | - | 1 - 2 | 238 | 62 | ||
| AK165581.1 | - | 1 - 2 | 259 | 62 | ||
| AK168888.1 | - | 1 - 2 | 232 | 62 | ||
| BC059239.1 | - | 1 - 2 | 218 | 6409279 | 62 | |
| BC060234.1 | - | 1 - 2 | 245 | 6853000 | 62 | |
| BC065102.1 | - | 1 - 2 | 235 | 6837587 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087706.1, AK133562.1, AK141750.1, AK151947.1, AK152309.1, AK161713.1, AK165581.1, AK168888.1, BC059239.1, BC060234.1, BC065102.1, NM_016714.2 transcripts.
Set Membership: |
|

Nup50
Genomic Map