Assay Details
| Gene Symbol: Nup50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016714.2 | NP_057923.2 | 2 - 3 | 297 | 80 | |
| GenBank mRNA | AF229256.1 | - | 1 - 2 | 64 | 80 | |
| AF251799.1 | - | 1 - 2 | 64 | 80 | ||
| AK087706.1 | - | 2 - 3 | 315 | 80 | ||
| AK133371.1 | - | 3 - 4 | 405 | 80 | ||
| AK133562.1 | - | 2 - 3 | 332 | 80 | ||
| AK141750.1 | - | 2 - 3 | 338 | 80 | ||
| AK151947.1 | - | 2 - 3 | 302 | 80 | ||
| AK152309.1 | - | 2 - 3 | 302 | 80 | ||
| AK161713.1 | - | 2 - 3 | 317 | 80 | ||
| AK165581.1 | - | 2 - 3 | 338 | 80 | ||
| AK168888.1 | - | 2 - 3 | 311 | 80 | ||
| BC059239.1 | - | 2 - 3 | 297 | 6409279 | 80 | |
| BC060234.1 | - | 2 - 3 | 324 | 6853000 | 80 | |
| BC065102.1 | - | 2 - 3 | 314 | 6837587 | 80 | |
More Information
Set Membership: |
|

Nup50
Genomic Map