Assay Details
| Gene Symbol: Nup50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016714.2 | NP_057923.2 | 6 - 7 | 1301 | 95 | |
| GenBank mRNA | AF229256.1 | - | 5 - 6 | 1068 | 95 | |
| AF251799.1 | - | 6 - 7 | 1068 | 95 | ||
| AK133562.1 | - | 6 - 7 | 1336 | 95 | ||
| AK141750.1 | - | 6 - 7 | 1342 | 95 | ||
| AK151947.1 | - | 6 - 7 | 1306 | 95 | ||
| AK152309.1 | - | 6 - 7 | 1306 | 95 | ||
| AK161713.1 | - | 6 - 7 | 1321 | 95 | ||
| AK165581.1 | - | 6 - 7 | 1342 | 95 | ||
| AK168888.1 | - | 6 - 7 | 1315 | 95 | ||
| BC059239.1 | - | 6 - 7 | 1301 | 6409279 | 95 | |
| BC060234.1 | - | 6 - 7 | 1328 | 6853000 | 95 | |
| BC065102.1 | - | 6 - 7 | 1318 | 6837587 | 95 | |
More Information
Set Membership: |
|

Nup50
Genomic Map