Assay Details
| Gene Symbol: Pdlim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016861.4 | NP_058557.2 | 3 - 4 | 508 | 76 | |
| GenBank mRNA | AF053367.2 | - | 3 - 4 | 424 | 76 | |
| AK076285.1 | - | 407 | 76 | |||
| AK088089.1 | - | 3 - 4 | 407 | 76 | ||
| AK146225.1 | - | 3 - 4 | 418 | 76 | ||
| AK146399.1 | - | 3 - 4 | 420 | 76 | ||
| AK158173.1 | - | 3 - 4 | 407 | 76 | ||
| BC004809.1 | - | 3 - 4 | 399 | 3588132 | 76 | |
More Information
Set Membership: |
|

Pdlim1
Genomic Map