Assay Details
| Gene Symbol: Insm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016889.3 | NP_058585.2 | 1 | 317 | 70 | |
| GenBank mRNA | AF044669.1 | - | 1 | 221 | 70 | |
| BC051240.1 | - | 1 | 393 | 6469481 | 70 | |
| BC082809.1 | - | 1 | 203 | 5720418 | 70 | |
More Information
Set Membership: |
|

Insm1
Genomic Map