Assay Details
| Gene Symbol: Gal3st1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016922.3 | NP_058618.2 | 2 - 3 | 420 | 89 | |
| GenBank mRNA | BC026806.1 | - | 2 - 3 | 343 | 4215616 | 89 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC026806.1, NM_016922.3 transcripts.
Set Membership: |
|

Gal3st1
Genomic Map