Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 13 - 14 | 2392 | 107 | |
| GenBank mRNA | AK076619.1 | - | 13 - 14 | 2390 | 107 | |
| BC026715.1 | - | 11 - 12 | 1851 | 107 | ||
| BC052672.1 | - | 13 - 14 | 2356 | 30058496 | 107 | |
More Information
Set Membership: |
|

Spag5
Genomic Map