Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 21 - 22 | 3345 | 97 | |
| GenBank mRNA | AF420307.1 | - | 21 - 22 | 3282 | 97 | |
| AK076619.1 | - | 21 - 22 | 3343 | 97 | ||
| BC052672.1 | - | 21 - 22 | 3309 | 30058496 | 97 | |
More Information
Set Membership: |
|

Spag5
Genomic Map