Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 22 - 23 | 3415 | 69 | |
| GenBank mRNA | AF420307.1 | - | 22 - 23 | 3352 | 69 | |
| AK076619.1 | - | 22 - 23 | 3413 | 69 | ||
More Information
Set Membership: |
|

Spag5
Genomic Map