Assay Details
| Gene Symbol: Spag5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017407.2 | NP_059103.1 | 2 - 3 | 242 | 98 | |
| GenBank mRNA | AF420307.1 | - | 2 - 3 | 179 | 98 | |
| AK052228.1 | - | 2 - 3 | 210 | 98 | ||
| AK076619.1 | - | 2 - 3 | 238 | 98 | ||
| BC052672.1 | - | 2 - 3 | 206 | 30058496 | 98 | |
| X81633.1 | - | 2 - 3 | 178 | 98 | ||
More Information
Set Membership: |
|

Spag5
Genomic Map