Assay Details
| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018732.3 | NP_061202.3 | 16 - 17 | 3052 | 106 | |
| GenBank mRNA | L42337.1 | - | 1 - 2 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn3a| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018732.3 | NP_061202.3 | 16 - 17 | 3052 | 106 | |
| GenBank mRNA | L42337.1 | - | 1 - 2 | 106 | ||
Set Membership: |
|