Assay Details
| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018732.3 | NP_061202.3 | 20 - 21 | 3977 | 85 | |
| GenBank mRNA | AK148712.1 | - | 2 - 3 | 1284 | 85 | |
| CD348085.1 | - | 1 - 2 | 172 | 85 | ||
More Information
Set Membership: |
|

Scn3a
Genomic Map