Assay Details
| Gene Symbol: Ramp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019511.3 | NP_062384.1 | 1 - 2 | 97 | 81 | |
| GenBank mRNA | AF146524.1 | - | 1 - 2 | 98 | 81 | |
| AF209907.1 | - | 1 - 2 | 57 | 81 | ||
| AJ250491.1 | - | 1 - 2 | 98 | 81 | ||
| BC024765.1 | - | 1 - 2 | 93 | 5364778 | 81 | |
More Information
Set Membership: |
|

Ramp3
Genomic Map