Assay Details
| Gene Symbol: Metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019648.3 | NP_062622.1 | 11 - 12 | 1872 | 117 | |
| GenBank mRNA | AB003144.1 | - | 10 - 11 | 1183 | 117 | |
| AF434712.1 | - | 10 - 11 | 1262 | 117 | ||
| AK053691.1 | - | 2 - 3 | 1253 | 117 | ||
| AK076804.1 | - | 10 - 11 | 1215 | 117 | ||
| AK090323.1 | - | 10 - 11 | 1201 | 117 | ||
| AK147097.1 | - | 10 - 11 | 1307 | 117 | ||
| AK159650.1 | - | 10 - 11 | 1286 | 117 | ||
| AK163318.1 | - | 10 - 11 | 1289 | 117 | ||
| AK165301.1 | - | 11 - 12 | 1874 | 117 | ||
| AK165845.1 | - | 10 - 11 | 1198 | 117 | ||
| BC002213.1 | - | 10 - 11 | 1284 | 3490341 | 117 | |
| BC010263.1 | - | 10 - 11 | 1256 | 117 | ||
| BC092052.1 | - | 10 - 11 | 1249 | 3670685 | 117 | |
More Information
Set Membership: |
|

Metap2
Genomic Map