Assay Details
| Gene Symbol: Kcnj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019659.3 | NP_062633.1 | 1 - 2 | 199 | 102 | |
| GenBank mRNA | AK085262.1 | - | 1 - 2 | 125 | 102 | |
| CF168846.1 | - | 1 - 2 | 92 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_019659.3 transcripts.
Set Membership: |
|

Kcnj1
Genomic Map