Assay Details
| Gene Symbol: Akap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019921.3 | NP_064305.2 | 10 - 11 | 1818 | 113 | |
| GenBank mRNA | AF021833.1 | - | 7 - 8 | 1172 | 113 | |
| AK049399.1 | - | 10 - 11 | 1787 | 113 | ||
| AK051044.1 | - | 8 - 9 | 1453 | 113 | ||
| BC038483.1 | - | 7 - 8 | 876 | 113 | ||
| BC054105.1 | - | 7 - 8 | 1121 | 30022728 | 113 | |
More Information
Set Membership: |
|

Akap10
Genomic Map