Assay Details
| Gene Symbol: Akap10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019921.3 | NP_064305.2 | 8 - 9 | 1499 | 92 | |
| GenBank mRNA | AF021833.1 | - | 5 - 6 | 853 | 92 | |
| AK049399.1 | - | 8 - 9 | 1468 | 92 | ||
| AK051044.1 | - | 6 - 7 | 1134 | 92 | ||
| AK083255.1 | - | 8 - 9 | 1468 | 92 | ||
| AK162966.1 | - | 6 - 7 | 1291 | 92 | ||
| BC038483.1 | - | 5 - 6 | 557 | 92 | ||
| BC054105.1 | - | 5 - 6 | 802 | 30022728 | 92 | |
More Information
Set Membership: |
|

Akap10
Genomic Map