Assay Details
| Gene Symbol: Cst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021405.2 | NP_067380.1 | 1 - 2 | 369 | 93 | |
| GenBank mRNA | AB036743.1 | - | 1 - 2 | 369 | 93 | |
| BC048364.1 | - | 1 - 2 | 285 | 4920219 | 93 | |
More Information
Set Membership: |
|

Cst10
Genomic Map