Assay Details
| Gene Symbol: Kcnmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021452.1 | NP_067427.1 | 1 - 2 | 336 | 79 | |
| GenBank mRNA | AF215892.1 | - | 1 - 2 | 336 | 79 | |
| AK013647.1 | - | 1 - 2 | 695 | 79 | ||
| BC048471.1 | - | 1 - 2 | 349 | 79 | ||
| BC117779.2 | - | 1 - 2 | 336 | 40091866 | 79 | |
More Information
Set Membership: |
|

Kcnmb4
Genomic Map