Assay Details
| Gene Symbol: Arhgap23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021493.2 | NP_067468.2 | 13 - 14 | 2767 | 88 | |
| GenBank mRNA | AK173187.1 | - | 4 - 5 | 413 | 88 | |
More Information
Set Membership: |
|

Arhgap23
Genomic Map