Assay Details
| Gene Symbol: Arhgap23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021493.2 | NP_067468.2 | 5 - 6 | 1687 | 54 | |
| GenBank mRNA | CB525670.1 | - | 3 - 4 | 230 | 54 | |
More Information
Set Membership: |
|

Arhgap23
Genomic Map