Assay Details
| Gene Symbol: Tcte2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022311.2 | NP_071706.2 | 1 - 2 | 113 | 121 | |
| GenBank mRNA | AK005879.1 | - | 98 | 121 | ||
| AK008572.1 | - | 99 | 121 | |||
| BC049646.1 | - | 1 - 2 | 105 | 121 | ||
| BC092528.1 | - | 1 - 2 | 113 | 6741782 | 121 | |
| U46151.1 | - | 1 - 2 | 320 | 121 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005879.1, AK008572.1, BC092528.1, NM_022311.2, U46151.1 transcripts.
Set Membership: |
|

Tcte2
Genomic Map