Assay Details
| Gene Symbol: Pbk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023209.2 | NP_075698.1 | 2 - 3 | 299 | 107 | |
| GenBank mRNA | AB041882.1 | - | 2 - 3 | 302 | 107 | |
| AK003838.1 | - | 2 - 3 | 260 | 107 | ||
| AK076121.1 | - | 2 - 3 | 372 | 107 | ||
| BC020099.1 | - | 2 - 3 | 305 | 3673060 | 107 | |
More Information
Set Membership: |
|

Pbk
Genomic Map