Assay Details
| Gene Symbol: Cdc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023223.2 | NP_075712.2 | 7 - 8 | 1301 | 96 | |
| GenBank mRNA | AB045313.1 | - | 7 - 8 | 1205 | 96 | |
| AF312208.1 | - | 7 - 8 | 1198 | 96 | ||
| AK011131.1 | - | 5 - 6 | 688 | 96 | ||
| AK011723.1 | - | 7 - 8 | 1300 | 96 | ||
| AK029424.1 | - | 7 - 8 | 1253 | 96 | ||
| AK075998.1 | - | 7 - 8 | 1249 | 96 | ||
| AK076030.1 | - | 7 - 8 | 1253 | 96 | ||
| AK083459.1 | - | 7 - 8 | 1130 | 96 | ||
| AK160204.1 | - | 7 - 8 | 1313 | 96 | ||
| AK168650.1 | - | 7 - 8 | 1253 | 96 | ||
| AK190077.1 | - | 78 | 96 | |||
| BC003215.1 | - | 7 - 8 | 1238 | 3157842 | 96 | |
More Information
Set Membership: |
|

Cdc20
Genomic Map