Assay Details
| Gene Symbol: Kbtbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034882.3 | NP_001030054.3 | 1 | 1222 | 102 | |
| GenBank mRNA | AK159684.1 | - | 1 | 1223 | 102 | |
| AK163295.1 | - | 2 | 1178 | 102 | ||
More Information
Set Membership: |
|

Kbtbd6
Genomic Map