Assay Details
| Gene Symbol: Prok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037539.2 | NP_001032628.1 | 1 - 2 | 162 | 84 | |
| NM_015768.2 | NP_056583.1 | 1 - 2 | 162 | 84 | ||
| GenBank mRNA | AF182064.1 | - | 1 - 2 | 162 | 84 | |
| AF182065.1 | - | 1 - 2 | 162 | 84 | ||
| AF182066.1 | - | 1 - 2 | 162 | 84 | ||
| AF487280.1 | - | 1 - 2 | 153 | 84 | ||
| AK015462.1 | - | 1 - 2 | 124 | 84 | ||
| BC116911.1 | - | 1 - 2 | 131 | 40126230 | 84 | |
| BC144863.1 | - | 1 - 2 | 143 | 9053411 | 84 | |
More Information
Set Membership: |
|

Prok2
Genomic Map