Assay Details
| Gene Symbol: Prok2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037539.2 | NP_001032628.1 | 2 - 3 | 293 | 86 | |
| GenBank mRNA | AF182066.1 | - | 2 - 3 | 293 | 86 | |
| AF487280.1 | - | 2 - 3 | 284 | 86 | ||
| BC144863.1 | - | 2 - 3 | 274 | 9053411 | 86 | |
| BY232101.1 | - | 2 - 3 | 309 | 86 | ||
More Information
Set Membership: |
|

Prok2
Genomic Map