Assay Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038635.2 | NP_001033724.2 | 2 - 3 | 936 | 90 | |
| GenBank mRNA | AK165988.1 | - | 2 - 3 | 831 | 90 | |
More Information
Set Membership: |
|

Stk35
Genomic Map