Assay Details
| Gene Symbol: Cyp4f18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024444.2 | NP_077764.2 | 2 - 3 | 246 | 79 | |
| GenBank mRNA | AF233647.1 | - | 2 - 3 | 237 | 79 | |
| AK007863.1 | - | 2 - 3 | 248 | 79 | ||
| BC013494.1 | - | 2 - 3 | 233 | 4218398 | 79 | |
More Information
Set Membership: |
|

Cyp4f18
Genomic Map