Assay Details
| Gene Symbol: Cyp4f18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024444.2 | NP_077764.2 | 3 - 4 | 389 | 106 | |
| GenBank mRNA | AF233647.1 | - | 3 - 4 | 380 | 106 | |
| AK007863.1 | - | 3 - 4 | 391 | 106 | ||
| BC013494.1 | - | 3 - 4 | 376 | 4218398 | 106 | |
More Information
Set Membership: |
|

Cyp4f18
Genomic Map