Assay Details
Gene Symbol: Stfa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025288.2 | NP_079564.1 | 2 - 3 | 220 | 83 | |
GenBank mRNA | AK014349.1 | - | 2 - 3 | 223 | 83 | |
AY169236.1 | - | 2 - 3 | 203 | 83 | ||
BC107224.1 | - | 2 - 3 | 214 | 40051891 | 83 | |
BC107225.1 | - | 2 - 3 | 216 | 40051892 | 83 | |
M92419.1 | - | 2 - 3 | 204 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
