Assay Details
| Gene Symbol: Trappc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025701.4 | NP_079977.3 | 1 - 2 | 131 | 83 | |
| GenBank mRNA | AK003633.1 | - | 1 - 2 | 99 | 83 | |
| AK007989.1 | - | 1 - 2 | 98 | 82 | ||
| AK076212.1 | - | 1 - 2 | 132 | 83 | ||
| AK078451.1 | - | 1 - 2 | 97 | 83 | ||
| BC049179.1 | - | 1 - 2 | 89 | 6313540 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003633.1, AK007989.1, AK076212.1, AK078451.1, BC049179.1, NM_025701.4 transcripts.
Set Membership: |
|

Trappc5
Genomic Map