Assay Details
| Gene Symbol: Cdca7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025866.3 | NP_080142.1 | 4 - 5 | 565 | 90 | |
| GenBank mRNA | AK011289.1 | - | 4 - 5 | 566 | 90 | |
| AK028671.1 | - | 4 - 5 | 576 | 90 | ||
| AK075882.1 | - | 4 - 5 | 530 | 90 | ||
| AK153943.1 | - | 4 - 5 | 544 | 90 | ||
| AK167734.1 | - | 4 - 5 | 566 | 90 | ||
| BC066169.1 | - | 4 - 5 | 573 | 30474357 | 90 | |
More Information
Set Membership: |
|

Cdca7
Genomic Map