Assay Details
| Gene Symbol: Atp5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025983.3 | NP_080259.1 | 2 - 3 | 262 | 93 | |
| GenBank mRNA | AK007752.1 | - | 2 - 3 | 251 | 93 | |
| AK007809.1 | - | 2 - 3 | 251 | 93 | ||
| AK010661.2 | - | 2 - 3 | 255 | 93 | ||
| AK012444.1 | - | 2 - 3 | 252 | 93 | ||
| AK131585.1 | - | 725 | 93 | |||
| AK160387.1 | - | 2 - 3 | 249 | 93 | ||
| AY341882.1 | - | 2 - 3 | 97 | |||
| BC024339.1 | - | 2 - 3 | 254 | 4981796 | 93 | |
| BC046612.1 | - | 1 - 2 | 1545 | 93 | ||
| CJ322223.1 | - | 2 - 3 | 262 | 93 | ||
| W78532.1 | - | 2 - 3 | 237 | 93 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ATP biosynthetic process
transport
ion transport
ATP synthesis coupled proton transport
proton transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
hydrolase activity
ATPase activity
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism
ATP biosynthetic process
transport
ion transport
ATP synthesis coupled proton transport
proton transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
hydrolase activity
ATPase activity
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875502 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875504 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875505 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875564 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875562 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P362054 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52703 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352966 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171399 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P362054 | - | - |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P362054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875552 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875563 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875558 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875556 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875560 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875568 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875557 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875564 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875540 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875569 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875566 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875567 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21875565 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171399 | - | - |
| Whole Mouse Genome G4122A | A_51_P362054 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127900 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171399 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01647847 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01647867 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01647863 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01647852 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01647860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875563 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875504 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875505 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875502 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875558 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875560 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875562 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21875564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875469 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21875640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02215258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02215268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02215275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01755945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01647855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875552 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01647845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01647843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875532 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875569 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21875567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01755948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10490224 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490222 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490223 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416567_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10490221 | - | - |
| Mouse Genome 430 2.0 Array | 1416567_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1416567_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67126_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67126_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200045328.mm.1 | NM_025983.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045328.mm.1 | NM_025983.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045328.mm.1 | TC0200005316.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045330.mm.1 | NM_025983.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045330.mm.1 | NM_025983.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045330.mm.1 | TC0200005316.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045333.mm.1 | NM_025983.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045333.mm.1 | NM_025983.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045333.mm.1 | TC0200005316.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1416567_s_at | - | - |
| Mouse Expression Set 430 | 1416567_PM_s_at | - | - |
| Murine Genome U74v2 Set | 93596_PM_i_at | - | - |
| Murine Genome U74v2 Set | 93596_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416567_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416567_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416567_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416567_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4120411 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2604794 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2604794 | - | - |
| mouse-6 v1.1 (Illumina) | scl18252.2_52-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120411 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18252.2_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120411 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2604794 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2604794 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl18252.2_52-S | - | - |

Atp5e
Genomic Map