Assay Details
| Gene Symbol: Hdhd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026108.3 | NP_080384.2 | 1 | 321 | 72 | |
| GenBank mRNA | AK007231.1 | - | 1 | 347 | 72 | |
| AK014922.1 | - | 1 | 298 | 72 | ||
| BC048447.1 | - | 1 | 310 | 6705877 | 72 | |
| CN834894.1 | - | 324 | 72 | |||
More Information
Set Membership: |
|

Hdhd1a
Genomic Map