Assay Details
| Gene Symbol: Ubl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026261.2 | NP_080537.1 | 1 | 419 | 88 | |
| GenBank mRNA | AK006408.1 | - | 1 | 345 | 88 | |
| AK015868.1 | - | 1 | 420 | 88 | ||
| AK016746.1 | - | 1 | 376 | 88 | ||
| BC048443.1 | - | 1 | 358 | 6775231 | 88 | |
| BC061071.1 | - | 1 | 384 | 6774419 | 88 | |
More Information
Set Membership: |
|

Ubl4b
Genomic Map